Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R2P9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002191.2 | 1 | 754 | |||
GenBank mRNA | AB044137.1 | - | 1 | 754 | ||
BC056673.1 | - | 1 | 684 |