Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LOC107983997 SIRNA Assays ›
          Gene Symbol:
          LOC107983997 ZNF83
          Gene Name:
          zinc finger protein 816-like
          zinc finger protein 83
          Gene Aliases:
          -
          HPF1, ZNF816B
          Chromosome Location:
          Chr.19: 52614711 - 52690527 on Build GRCh38
          Chromosome Location:
          Chr.19: 52612364 - 52690581 on Build GRCh38
          UniGene ID:
          -
          Hs.467210
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID n268878

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LOC107983997
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_011527096.2 XP_011525398.1 2 118
          Gene Symbol: ZNF83
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001277945.1 NP_001264874.1 2 172
          NM_001277946.1 NP_001264875.1 2 172
          NM_001277947.1 NP_001264876.1 3 244
          NM_001277948.1 NP_001264877.1 2 172
          NM_001277949.1 NP_001264878.1 2 172
          XM_017026951.1 XP_016882440.1 2 150
          GenBank mRNA AK075066.1 - 2 79
          AK289399.1 - 2 77
          BG399319.1 - 2 173
          DA434207.1 - 2 77
          DA701045.1 - 3 149
          DB134270.1 - 2 82
          DB254966.1 - 2 78
          DB272526.1 - 2 82
          DT216882.1 - 3 158
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DR001735.1 - 2 123 Chr. 19 - 52618790 - 52690526 [-] on Build GRCh38
          GenBankmRNA HY058399.1 - 2 94 Chr. 19 - 52618928 - 52690502 [-] on Build GRCh38
          GenBankmRNA DA795304.1 - 2 134 Chr. 19 - 52614770 - 52690543 [-] on Build GRCh38
          GenBankmRNA DA237809.1 - 2 106 Chr. 19 - 52654102 - 52690515 [-] on Build GRCh38
          GenBankmRNA DA023799.1 - 2 77 Chr. 19 - 52614466 - 52690486 [-] on Build GRCh38
          GenBankmRNA DA067188.1 - 2 82 Chr. 19 - 52654097 - 52690491 [-] on Build GRCh38
          GenBankmRNA DA863416.1 - 2 79 Chr. 19 - 52617248 - 52690488 [-] on Build GRCh38
          GenBankmRNA DA828340.1 - 2 79 Chr. 19 - 52614521 - 52690488 [-] on Build GRCh38
          GenBankmRNA DA626221.1 - 2 87 Chr. 19 - 52654116 - 52690496 [-] on Build GRCh38
          GenBankmRNA DB238373.1 - 2 87 Chr. 19 - 52654071 - 52690496 [-] on Build GRCh38
          GenBankmRNA DA108951.1 - 2 87 Chr. 19 - 52617358 - 52690496 [-] on Build GRCh38
          GenBankmRNA DB156129.1 - 2 77 Chr. 19 - 52616717 - 52690486 [-] on Build GRCh38
          GenBankmRNA DA108939.1 - 2 87 Chr. 19 - 52617292 - 52690496 [-] on Build GRCh38
          GenBankmRNA AV764447.1 - 75 Chr. - - [-] on Build
          GenBankmRNA DA924899.1 - 2 83 Chr. 19 - 52614690 - 52690492 [-] on Build GRCh38
          GenBankmRNA DA106367.1 - 2 79 Chr. 19 - 52618699 - 52690488 [-] on Build GRCh38
          GenBankmRNA DA684911.1 - 2 78 Chr. 19 - 52614716 - 52690486 [-] on Build GRCh38
          GenBankmRNA DA406119.1 - 2 131 Chr. 19 - 52617107 - 52690540 [-] on Build GRCh38
          GenBankmRNA DA125855.1 - 2 115 Chr. 19 - 52617246 - 52690523 [-] on Build GRCh38
          GenBankmRNA DA231238.1 - 2 87 Chr. 19 - 52618869 - 52690496 [-] on Build GRCh38
          GenBankmRNA DW451571.1 - 2 88 Chr. 19 - 52655451 - 52690480 [-] on Build GRCh38
          GenBankmRNA DA397058.1 - 2 78 Chr. 19 - 52614680 - 52690486 [-] on Build GRCh38
          GenBankmRNA DA109862.1 - 2 79 Chr. 19 - 52618675 - 52690488 [-] on Build GRCh38
          GenBankmRNA DA100765.1 - 2 87 Chr. 19 - 52618712 - 52690496 [-] on Build GRCh38
          GenBankmRNA DA589517.1 - 2 78 Chr. 19 - 52614606 - 52690486 [-] on Build GRCh38
          GenBankmRNA DB095489.1 - 2 150 Chr. 19 - 52618934 - 52690559 [-] on Build GRCh38
          GenBankmRNA AL583521.3 - 1 34 Chr. 19 - 52594079 - 52660844 [-] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0