Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MIR100HG SIRNA Assays ›
          Gene Symbol:
          MIR100HG
          Gene Name:
          mir-100-let-7a-2 cluster host gene
          Gene Aliases:
          AGD1, linc-NeD125, lncRNA-N2
          Chromosome Location:
          Chr.11: 122089103 - 122203062 on Build GRCh38
          UniGene ID:
          Hs.44098
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n269148

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MIR100HG
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_024430.2 4 1973
          NR_137175.1 2 2244
          NR_137176.1 2 1955
          NR_137177.1 2 1934
          NR_137179.1 5 1927
          NR_137180.1 5 1985
          NR_137181.1 3 1870
          NR_137182.1 4 1952
          NR_137183.1 3 1861
          NR_137184.1 2 1779
          NR_137185.1 3 1840
          NR_137186.1 3 1749
          NR_137187.1 3 1728
          NR_137188.1 3 1818
          NR_137189.1 2 2265
          NR_137190.1 3 2298
          NR_137191.1 3 2277
          NR_137192.1 4 2137
          NR_137193.1 5 1964
          NR_137194.1 3 1839
          NR_137195.1 2 2007
          NR_137196.1 2 2028
          GenBank mRNA AB075498.1 - 2 2497
          AK021552.1 - 1 1139
          AK123947.1 - 2 2181
          AL832469.1 - 3 1533
          BC040599.1 - 2 1557
          BX538058.1 - 2 1535
          BX647608.1 - 2 1535
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DT217205.1 - 1 187 Chr. 11 - 122089813 - 122090445 [-] on Build GRCh38
          GenBankmRNA H17681.1 - 1 303 Chr. 11 - 122090077 - 122090561 [-] on Build GRCh38
          GenBankmRNA AW160705.1 - 1 387 Chr. 11 - 122090206 - 122090639 [-] on Build GRCh38
          GenBankmRNA DA771923.1 - 1 77 Chr. 11 - 122089739 - 122090335 [-] on Build GRCh38
          GenBankmRNA CV570306.1 - 1 261 Chr. 11 - 122089878 - 122090519 [-] on Build GRCh38
          GenBankmRNA BM541657.1 - 1 178 Chr. 11 - 122089494 - 122090435 [-] on Build GRCh38
          GenBankmRNA CN345412.1 - 1 32 Chr. 11 - 122089688 - 122090290 [-] on Build GRCh38
          GenBankmRNA BQ888152.1 - 121 Chr. - - [-] on Build
          GenBankmRNA AU139742.1 - 1 377 Chr. 11 - 122089958 - 122090635 [-] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0