Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIO3OS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002770.1 | 1 | 2800 | |||
GenBank mRNA | AF469201.1 | - | 2 | 1819 | ||
AF469202.1 | - | 1 | 2800 |