Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RNF216P1 SIRNA Assays ›
          Gene Symbol:
          RNF216P1
          Gene Name:
          ring finger protein 216 pseudogene 1
          Gene Aliases:
          RNF216L, hCG2040556
          Chromosome Location:
          Chr.7: 4973985 - 4998169 on Build GRCh38
          UniGene ID:
          Hs.520636
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n269571

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RNF216P1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_015449.1 6 752
          NR_023384.1 8 925
          NR_023385.1 5 637
          GenBank mRNA AK124529.1 - 2 2130
          AK303831.1 - 5 488
          AK308526.1 - 5 572
          AK309798.1 - 5 496
          BC026736.1 - 6 631
          BC068459.1 - 8 894
          BX538059.1 - 6 679
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BU187037.1 - 6 664 Chr. 7 - 4974073 - 4996911 [+] on Build GRCh38
          GenBankmRNA R18687.1 - 2 142 Chr. 7 - 4995496 - 4997034 [+] on Build GRCh38
          GenBankmRNA AL520766.3 - 6 687 Chr. 7 - 4974050 - 4996915 [+] on Build GRCh38
          GenBankmRNA DC413548.1 - 5 488 Chr. 7 - 4974082 - 4996763 [+] on Build GRCh38
          GenBankmRNA BI916700.1 - 6 673 Chr. 7 - 4976312 - 4996882 [+] on Build GRCh38
          GenBankmRNA DR159194.1 - 6 677 Chr. 7 - 4974072 - 4996813 [+] on Build GRCh38
          GenBankmRNA BG120560.1 - 2 148 Chr. 7 - 4995490 - 4997223 [+] on Build GRCh38
          GenBankmRNA F11246.1 - 143 Chr. - - [-] on Build
          GenBankmRNA AL530709.3 - 3 280 Chr. 7 - 4989061 - 4997497 [+] on Build GRCh38
          GenBankmRNA DC412017.1 - 5 515 Chr. 7 - 4974056 - 4996725 [+] on Build GRCh38
          GenBankmRNA DC340388.1 - 2 415 Chr. 7 - 4984716 - 4996774 [+] on Build GRCh38
          GenBankmRNA AL521852.3 - 6 700 Chr. 7 - 4974037 - 4996882 [+] on Build GRCh38
          GenBankmRNA BM702294.1 - 2 196 Chr. 7 - 4995442 - 4997180 [+] on Build GRCh38
          GenBankmRNA AA214166.1 - 3 238 Chr. 7 - 4989104 - 4996811 [+] on Build GRCh38
          GenBankmRNA BM805546.1 - 7 738 Chr. 7 - 4974067 - 4996987 [+] on Build GRCh38
          GenBankmRNA BQ881985.1 - 481 Chr. - - [-] on Build
          GenBankmRNA BQ434583.1 - 7 746 Chr. 7 - 4974115 - 4996822 [+] on Build GRCh38
          GenBankmRNA BE303029.1 - 5 496 Chr. 7 - 4983722 - 4996754 [+] on Build GRCh38
          GenBankmRNA BI871291.1 - 4 445 Chr. 7 - 4983695 - 4997114 [+] on Build GRCh38
          GenBankmRNA HY149305.1 - 2 126 Chr. 7 - 4995512 - 4997022 [+] on Build GRCh38
          GenBankmRNA DN998801.1 - 6 678 Chr. 7 - 4974059 - 4996695 [+] on Build GRCh38
          GenBankmRNA BG024063.1 - 210 Chr. - - [-] on Build
          GenBankmRNA DA696204.1 - 2 375 Chr. 7 - 4995264 - 4996851 [+] on Build GRCh38
          GenBankmRNA DC301966.1 - 5 573 Chr. 7 - 4974050 - 4996693 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0