Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR2A9P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002157.1 | 1 | 855 |
Gene Symbol: OR2A20P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK172840.1 | - | 1 | 2357 | ||
BC016940.1 | - | 3 | 1431 |