Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NUTM2A-AS1 SIRNA Assays ›
          Gene Symbol:
          NUTM2A-AS1 NUTM2B-AS1
          Gene Name:
          NUTM2A antisense RNA 1
          NUTM2B antisense RNA 1
          Gene Aliases:
          FAM22A-AS1
          -
          Chromosome Location:
          Chr.10: 87238667 - 87342558 on Build GRCh38
          Chromosome Location:
          Chr.10: 79691500 - 79826594 on Build GRCh38
          UniGene ID:
          Hs.440648
          Hs.440648
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID n270296

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NUTM2A-AS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_024397.1 2 403
          GenBank mRNA AK309777.1 - 2 222
          AK311174.1 - 2 242
          BC047063.1 - 2 330
          DB257428.1 - 2 404
          Gene Symbol: NUTM2B-AS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_120611.1 2 483
          NR_120612.1 2 483
          NR_120613.1 2 483
          GenBank mRNA BQ425131.1 - 2 503
          CB163469.1 - 2 300
          CN279211.1 - 2 343
          CN279213.1 - 2 327
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DA040049.1 - 2 222 Chr. 10 - 87245755 - 87342377 [-] on Build GRCh38
          GenBankmRNA DA737563.1 - 2 353 Chr. 10 - 87245656 - 87342506 [-] on Build GRCh38
          GenBankmRNA BI767196.1 - 211 Chr. - - [-] on Build
          GenBankmRNA DA796822.1 - 2 387 Chr. 10 - 87288387 - 87342541 [-] on Build GRCh38
          GenBankmRNA AL520958.3 - 2 126 Chr. 10 - 79797377 - 79826237 [-] on Build GRCh38
          GenBankmRNA DA691562.1 - 2 366 Chr. 10 - 79804494 - 79826468 [-] on Build GRCh38
          GenBankmRNA BG111204.1 - 2 144 Chr. 10 - 79691671 - 79826255 [-] on Build GRCh38
          GenBankmRNA DA010830.1 - 3 417 Chr. 10 - 79806056 - 79826563 [-] on Build GRCh38
          GenBankmRNA BG166579.1 - 2 382 Chr. 10 - 79766137 - 79826484 [-] on Build GRCh38
          GenBankmRNA CD250242.1 - 2 410 Chr. 10 - 87288178 - 87342541 [-] on Build GRCh38
          GenBankmRNA HY154971.1 - 2 331 Chr. 10 - 87216096 - 87342485 [-] on Build GRCh38
          GenBankmRNA CN274008.1 - 2 215 Chr. 10 - 79802853 - 79826326 [-] on Build GRCh38
          GenBankmRNA BM559658.1 - 319 Chr. - - [-] on Build
          GenBankmRNA HY062022.1 - 2 279 Chr. 10 - 87288324 - 87342412 [-] on Build GRCh38
          GenBankmRNA DB483325.1 - 2 301 Chr. 10 - 87246167 - 87342456 [-] on Build GRCh38
          GenBankmRNA BG437437.1 - 451 Chr. - - [-] on Build
          GenBankmRNA HY087386.1 - 2 279 Chr. 10 - 87288299 - 87342412 [-] on Build GRCh38
          GenBankmRNA DA200463.1 - 2 242 Chr. 10 - 87206423 - 87342397 [-] on Build GRCh38
          GenBankmRNA CN279215.1 - 2 367 Chr. 10 - 79804484 - 79826470 [-] on Build GRCh38
          GenBankmRNA AW612231.1 - 2 281 Chr. 10 - 79804492 - 79826392 [-] on Build GRCh38
          GenBankmRNA HY150000.1 - 2 221 Chr. 10 - 87216123 - 87342375 [-] on Build GRCh38
          GenBankmRNA T58826.1 - 138 Chr. - - [-] on Build
          GenBankmRNA HY010169.1 - 2 301 Chr. 10 - 87238758 - 87342455 [-] on Build GRCh38
          GenBankmRNA HY124799.1 - 2 193 Chr. 10 - 87216064 - 87342347 [-] on Build GRCh38
          GenBankmRNA BG748770.1 - 2 405 Chr. 10 - 79766290 - 79826477 [-] on Build GRCh38
          GenBankmRNA BE742363.1 - 300 Chr. - - [-] on Build
          GenBankmRNA DB456680.1 - 2 330 Chr. 10 - 87246163 - 87342485 [-] on Build GRCh38
          GenBankmRNA HY151352.1 - 2 323 Chr. 10 - 87208482 - 87342477 [-] on Build GRCh38
          GenBankmRNA DA571282.1 - 2 245 Chr. 10 - 87288087 - 87342400 [-] on Build GRCh38
          GenBankmRNA DA829196.1 - 2 200 Chr. 10 - 87287993 - 87342355 [-] on Build GRCh38
          GenBankmRNA DA258253.1 - 2 222 Chr. 10 - 87216011 - 87342377 [-] on Build GRCh38
          GenBankmRNA DA259512.1 - 2 228 Chr. 10 - 87202973 - 87342383 [-] on Build GRCh38
          GenBankmRNA DA551755.1 - 2 188 Chr. 10 - 87245749 - 87342343 [-] on Build GRCh38
          GenBankmRNA BG717953.1 - 2 418 Chr. 10 - 79691997 - 79826516 [-] on Build GRCh38
          GenBankmRNA DB263965.1 - 2 215 Chr. 10 - 87206378 - 87342370 [-] on Build GRCh38
          GenBankmRNA HY055033.1 - 2 301 Chr. 10 - 87288436 - 87342455 [-] on Build GRCh38
          GenBankmRNA DA009741.1 - 2 220 Chr. 10 - 87245758 - 87342375 [-] on Build GRCh38
          GenBankmRNA BM053142.1 - 2 413 Chr. 10 - 79808759 - 79826515 [-] on Build GRCh38
          GenBankmRNA BF437950.1 - 2 230 Chr. 10 - 79801984 - 79826341 [-] on Build GRCh38
          GenBankmRNA DA775058.1 - 2 223 Chr. 10 - 87215939 - 87342377 [-] on Build GRCh38
          GenBankmRNA HY157097.1 - 2 193 Chr. 10 - 87216111 - 87342347 [-] on Build GRCh38
          GenBankmRNA BF223885.1 - 1 38 Chr. 10 - 79797859 - 79814658 [-] on Build GRCh38
          GenBankmRNA BM456581.1 - 626 Chr. - - [-] on Build
          GenBankmRNA BG256210.1 - 272 Chr. - - [-] on Build
          GenBankmRNA CN279212.1 - 2 325 Chr. 10 - 79804510 - 79826427 [-] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0