Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC105369199 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011542532.2 | XP_011540834.1 | 5 | 2169 | ||
XM_011542534.2 | XP_011540836.1 | 5 | 2149 | |||
XM_017003016.1 | XP_016858505.1 | 5 | 2097 |