Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: URAHP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027335.2 | 3 | 291 | |||
NR_027336.2 | 3 | 244 | ||||
GenBank mRNA | AK304171.1 | - | 3 | 234 | ||
BC044232.1 | - | 3 | 291 | |||
BC089450.1 | - | 3 | 227 | |||
DC393803.1 | - | 3 | 245 |
Gene Symbol: GAS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC036781.1 | - | 1 | 314 |