Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other JMJD4 SIRNA Assays ›
          Gene Symbol:
          JMJD4 SNAP47
          Gene Name:
          jumonji domain containing 4
          synaptosome associated protein 47
          Gene Aliases:
          -
          C1orf142, ESFI5812, HEL-S-290, HEL170, SNAP-47, SVAP1
          Chromosome Location:
          Chr.1: 227731189 - 227735423 on Build GRCh38
          Chromosome Location:
          Chr.1: 227728168 - 227781231 on Build GRCh38
          UniGene ID:
          Hs.555974
          Hs.325081
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID n270671

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: JMJD4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC109272.2 - 1 1096
          Gene Symbol: SNAP47
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_017000231.1 XP_016855720.1 3 588
          XM_017000232.1 XP_016855721.1 2 394
          GenBank mRNA AK055266.1 - 3 567
          AK093736.1 - 2 397
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DA626209.1 - 3 459 Chr. 1 - 227728647 - 227732623 [+] on Build GRCh38
          GenBankmRNA DB172854.1 - 3 462 Chr. 1 - 227728644 - 227732655 [+] on Build GRCh38
          GenBankmRNA DA046891.1 - 3 459 Chr. 1 - 227728647 - 227732821 [+] on Build GRCh38
          GenBankmRNA DA047186.1 - 3 420 Chr. 1 - 227728686 - 227732698 [+] on Build GRCh38
          GenBankmRNA DA928797.1 - 3 447 Chr. 1 - 227728661 - 227732642 [+] on Build GRCh38
          GenBankmRNA DA689849.1 - 3 433 Chr. 1 - 227728673 - 227732650 [+] on Build GRCh38
          GenBankmRNA DA197678.1 - 3 420 Chr. 1 - 227728687 - 227732684 [+] on Build GRCh38
          GenBankmRNA DA050036.1 - 2 257 Chr. 1 - 227728686 - 227732749 [+] on Build GRCh38
          GenBankmRNA DA360441.1 - 3 455 Chr. 1 - 227728651 - 227732632 [+] on Build GRCh38
          GenBankmRNA DA629296.1 - 3 457 Chr. 1 - 227728651 - 227732668 [+] on Build GRCh38
          GenBankmRNA DA878521.1 - 3 410 Chr. 1 - 227728696 - 227732706 [+] on Build GRCh38
          GenBankmRNA DA751189.1 - 3 468 Chr. 1 - 227728638 - 227732636 [+] on Build GRCh38
          GenBankmRNA DA706865.1 - 3 455 Chr. 1 - 227728651 - 227732665 [+] on Build GRCh38
          GenBankmRNA DA954484.1 - 2 333 Chr. 1 - 227728610 - 227732764 [+] on Build GRCh38
          GenBankmRNA DB103578.1 - 3 476 Chr. 1 - 227728630 - 227732629 [+] on Build GRCh38
          GenBankmRNA DA053725.1 - 2 257 Chr. 1 - 227728686 - 227732843 [+] on Build GRCh38
          GenBankmRNA DA716543.1 - 3 420 Chr. 1 - 227728686 - 227732669 [+] on Build GRCh38
          GenBankmRNA DA526492.1 - 4 533 Chr. 1 - 227728168 - 227732582 [+] on Build GRCh38
          GenBankmRNA DA873744.1 - 3 455 Chr. 1 - 227728651 - 227732660 [+] on Build GRCh38
          GenBankmRNA DB108655.1 - 2 397 Chr. 1 - 227728549 - 227732723 [+] on Build GRCh38
          GenBankmRNA DA497385.1 - 3 475 Chr. 1 - 227728631 - 227732585 [+] on Build GRCh38
          GenBankmRNA DA176792.1 - 2 324 Chr. 1 - 227728619 - 227732787 [+] on Build GRCh38
          GenBankmRNA DA634770.1 - 3 455 Chr. 1 - 227728651 - 227732649 [+] on Build GRCh38
          GenBankmRNA DA917153.1 - 3 420 Chr. 1 - 227728686 - 227732668 [+] on Build GRCh38
          GenBankmRNA DA509529.1 - 3 455 Chr. 1 - 227728651 - 227732642 [+] on Build GRCh38
          GenBankmRNA DA177526.1 - 3 470 Chr. 1 - 227728636 - 227732639 [+] on Build GRCh38
          GenBankmRNA DA796130.1 - 3 463 Chr. 1 - 227728647 - 227732607 [+] on Build GRCh38
          GenBankmRNA DA926452.1 - 3 420 Chr. 1 - 227728686 - 227732693 [+] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          SNARE protein

          Gene Ontology Categories:

          Function(s) Process(es)

          synaptic vesicle priming
          synaptic vesicle fusion to presynaptic active zone membrane
          long-term synaptic potentiation
          SNAP receptor activity
          protein binding
          syntaxin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0