Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ANKRD19P SIRNA Assays ›
          Gene Symbol:
          ANKRD19P EEF1D
          Gene Name:
          ankyrin repeat domain 19, pseudogene
          eukaryotic translation elongation factor 1 delta
          Gene Aliases:
          ANKRD19, bA526D8.2
          EF-1D, EF1D, FP1047
          Chromosome Location:
          Chr.9: 92809611 - 92838457 on Build GRCh38
          Chromosome Location:
          Chr.8: 143579697 - 143597675 on Build GRCh38
          UniGene ID:
          Hs.643597
          Hs.333388
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID n271005

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ANKRD19P
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_026868.1 9 1216
          GenBank mRNA AK093497.1 - 7 808
          BC028712.1 - 9 1180
          BC038951.1 - 8 990 5733799
          Gene Symbol: EEF1D
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC046445.1 - 1 1460
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA HY251851.1 - 1 119 Chr. 8 - 143579728 - 143580705 [+] on Build GRCh38
          GenBankmRNA AL041075.1 - 2 124 Chr. 9 - 92829114 - 92837198 [+] on Build GRCh38
          GenBankmRNA HY325640.1 - 1 119 Chr. 8 - 143579728 - 143580648 [+] on Build GRCh38
          GenBankmRNA HY374423.1 - 1 121 Chr. 8 - 143579728 - 143580700 [+] on Build GRCh38
          GenBankmRNA HY340883.1 - 1 117 Chr. 8 - 143579728 - 143580617 [+] on Build GRCh38
          GenBankmRNA HY262095.1 - 1 120 Chr. 8 - 143579728 - 143580668 [+] on Build GRCh38
          GenBankmRNA HY327126.1 - 1 118 Chr. 8 - 143579728 - 143580611 [+] on Build GRCh38
          GenBankmRNA HY310037.1 - 1 120 Chr. 8 - 143579727 - 143580651 [+] on Build GRCh38
          GenBankmRNA HY279031.1 - 1 119 Chr. 8 - 143579728 - 143580604 [+] on Build GRCh38
          GenBankmRNA HY355872.1 - 1 121 Chr. 8 - 143579728 - 143580659 [+] on Build GRCh38
          GenBankmRNA HY290035.1 - 1 121 Chr. 8 - 143579728 - 143580634 [+] on Build GRCh38
          GenBankmRNA HY292533.1 - 1 118 Chr. 8 - 143579728 - 143580642 [+] on Build GRCh38
          GenBankmRNA HY263668.1 - 1 120 Chr. 8 - 143579728 - 143580664 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-nglt7:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0