siRNA Details
Gene Symbol: LINC00964 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027321.1 | 2 | 3060 | |||
GenBank mRNA | AK056899.1 | - | 1 | 1434 | ||
BC043249.2 | - | 2 | 3013 |

Gene Symbol: LINC00964 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027321.1 | 2 | 3060 | |||
GenBank mRNA | AK056899.1 | - | 1 | 1434 | ||
BC043249.2 | - | 2 | 3013 |