Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282275.1 | NP_001269204.1 | 2 | 536 | ||
NM_001282276.1 | NP_001269205.1 | 2 | 470 | |||
XM_006721865.3 | XP_006721928.1 | 2 | 1152 | |||
XM_011524747.2 | XP_011523049.1 | 2 | 462 | |||
XM_011524748.1 | XP_011523050.1 | 2 | 407 | |||
GenBank mRNA | AF532777.1 | - | 2 | 524 | ||
BX106051.1 | - | 2 | 470 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CB048981.1 | - | 2 | 500 | Chr. 17 - 48723209 - 48724729 [+] on Build GRCh38 |