Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNORA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002920.1 | 1 | 14 | |||
GenBank mRNA | AJ609454.1 | - | 1 | 14 |
Gene Symbol: TAF1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK057183.1 | - | 1 | 355 | ||
AK128061.1 | - | 1 | 3363 |