Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIR3DX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026716.2 | 3, 4 | 458 | |||
NR_136268.1 | 3, 4 | 458 | ||||
GenBank mRNA | BC171834.1 | - | 2, 3 | 668 | ||
DQ224421.1 | - | 3, 4 | 359 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DN998408.1 | - | 1, 2 | 227 | Chr. 19 - 54533794 - 54536885 [+] on Build GRCh38 |