Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOTAIR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003716.3 | 3 | 285 | |||
NR_047517.1 | 4 | 344 | ||||
NR_047518.1 | 2 | 317 | ||||
GenBank mRNA | AK123741.1 | - | 2 | 98 | ||
DA567507.1 | - | 3 | 260 | |||
DA905601.1 | - | 4 | 344 | |||
DA907429.1 | - | 2 | 318 | |||
DQ926657.1 | - | 3 | 205 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA907474.1 | - | 2 | 318 | Chr. 12 - 53964096 - 53968914 [-] on Build GRCh38 |