Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_015379.3 | 3 | 921 | |||
GenBank mRNA | AK024303.1 | - | 2 | 1101 | ||
AK092401.1 | - | 3 | 907 | |||
BC005351.1 | - | 3 | 773 | |||
BC017567.1 | - | 3 | 777 | |||
BC107722.1 | - | 3 | 777 | |||
DQ343132.1 | - | LIT3336 UCA1 |
3 | 921 | ||
EU334869.1 | - | 3 | 922 | |||
GU357550.1 | - | 3 | 773 | |||
GU799561.1 | - | 3 | 921 | |||
GU799562.1 | - | 3 | 921 | |||
GU799563.1 | - | 3 | 921 | |||
GU799564.1 | - | 3 | 921 | |||
GU799565.1 | - | 3 | 921 | |||
HQ833208.1 | - | 3 | 921 | |||
HQ833209.1 | - | 3 | 921 |