Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL19P12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026660.1 | 1 | 326 | |||
GenBank mRNA | AW373666.1 | - | 1 | 328 | ||
CD709306.1 | - | 1 | 329 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AW373669.1 | - | 1 | 326 | Chr. 7 - 103141771 - 103142399 [-] on Build GRCh38 |