siRNA Details
Gene Symbol: CDK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AY927560.1 | - | LIT3563 AY927560 |
4 | 637 |

Gene Symbol: CDK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AY927560.1 | - | LIT3563 AY927560 |
4 | 637 |