siRNA Details
Gene Symbol: DSTNP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | U72518.1 | - | LIT2102 destrin-2 pseudogene |
1 | 457 |

Gene Symbol: DSTNP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | U72518.1 | - | LIT2102 destrin-2 pseudogene |
1 | 457 |