Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEATR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005257570.3 | XP_005257627.1 | 21 | 5291 | ||
XM_011525110.2 | XP_011523412.1 | 19 | 4990 | |||
XM_011525111.2 | XP_011523413.1 | 19 | 4952 | |||
XM_017024936.1 | XP_016880425.1 | 20 | 5364 | |||
XM_017024937.1 | XP_016880426.1 | 20 | 7040 | |||
XM_017024938.1 | XP_016880427.1 | 20 | 5096 | |||
XM_017024939.1 | XP_016880428.1 | 20 | 5367 | |||
GenBank mRNA | BC040714.1 | - | 1 | 968 |