siRNA Details
Gene Symbol: TPBG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011536097.2 | XP_011534399.1 | 2 | 6242 | ||
GenBank mRNA | AJ420536.1 | - | 1 | 1323 |

Gene Symbol: TPBG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011536097.2 | XP_011534399.1 | 2 | 6242 | ||
GenBank mRNA | AJ420536.1 | - | 1 | 1323 |