Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ID4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001546.3 | NP_001537.1 | 3 | 2467 | ||
GenBank mRNA | AJ420553.1 | - | 1 | 719 |