siRNA Details
Gene Symbol: CDYL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152342.3 | NP_689555.2 | 7 | 6374 | ||
XM_011522866.1 | XP_011521168.1 | 7 | 6386 | |||
XM_011522867.2 | XP_011521169.1 | 7 | 6929 | |||
GenBank mRNA | AK023660.1 | - | 1 | 307 |

More Information
Panther Classification:
Molecular Function -










