Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SPTLC2 SIRNA Assays ›
          Gene Symbol:
          SPTLC2
          Gene Name:
          serine palmitoyltransferase long chain base subunit 2
          Gene Aliases:
          HSN1C, LCB2, LCB2A, NSAN1C, SPT2, hLCB2a
          Chromosome Location:
          Chr.14: 77505997 - 77616767 on Build GRCh38
          UniGene ID:
          Hs.435661
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n284861

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SPTLC2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_004863.3 NP_004854.1 12 7811
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BF922779.1 - 1 121 Chr. 14 - 77506081 - 77506463 [-] on Build GRCh38
          GenBankmRNA CR616151.1 - 1 453 Chr. 14 - 77506038 - 77506802 [-] on Build GRCh38
          GenBankmRNA BI056671.1 - 1 413 Chr. 14 - 77506141 - 77506760 [-] on Build GRCh38
          GenBankmRNA BE827655.1 - 1 159 Chr. 14 - 77506287 - 77506509 [-] on Build GRCh38
          GenBankmRNA BE967498.1 - 1 260 Chr. 14 - 77506230 - 77506609 [-] on Build GRCh38
          GenBankmRNA AA256622.1 - 1 179 Chr. 14 - 77506176 - 77506528 [-] on Build GRCh38
          GenBankmRNA BF910355.1 - 1 367 Chr. 14 - 77506234 - 77506697 [-] on Build GRCh38
          GenBankmRNA AW996574.1 - 1 328 Chr. 14 - 77506287 - 77506677 [-] on Build GRCh38
          GenBankmRNA AA235169.1 - 1 179 Chr. 14 - 77506080 - 77506528 [-] on Build GRCh38
          GenBankmRNA BE827681.1 - 1 124 Chr. 14 - 77506293 - 77506444 [-] on Build GRCh38
          GenBankmRNA N92160.1 - 1 309 Chr. 14 - 77506159 - 77506658 [-] on Build GRCh38
          GenBankmRNA BF795943.1 - 1 352 Chr. 14 - 77506011 - 77506701 [-] on Build GRCh38
          GenBankmRNA CN430770.1 - 1 643 Chr. 14 - 77506269 - 77506990 [-] on Build GRCh38
          GenBankmRNA BQ325944.1 - 1 329 Chr. 14 - 77506281 - 77506664 [-] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transaminase

          Gene Ontology Categories:

          Function(s) Process(es)

          sphingomyelin biosynthetic process
          sphingolipid biosynthetic process
          sphinganine biosynthetic process
          sphingosine biosynthetic process
          ceramide biosynthetic process
          positive regulation of lipophagy
          serine C-palmitoyltransferase activity
          pyridoxal phosphate binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-xq6xf:80/100.66.76.183:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0