Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MAGI1 SIRNA Assays ›
          Gene Symbol:
          MAGI1
          Gene Name:
          membrane associated guanylate kinase, WW and PDZ domain containing 1
          Gene Aliases:
          AIP-3, AIP3, BAIAP1, BAP-1, BAP1, MAGI-1, Magi1d, TNRC19, WWP3
          Chromosome Location:
          Chr.3: 65353525 - 66038836 on Build GRCh38
          UniGene ID:
          Hs.476636
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n288421

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MAGI1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_005265563.1 XP_005265620.1 24 7208
          XM_005265564.1 XP_005265621.1 24 7205
          XM_005265565.1 XP_005265622.1 24 7175
          XM_005265566.1 XP_005265623.1 24 7169
          XM_005265568.1 XP_005265625.1 23 7166
          XM_005265570.1 XP_005265627.1 23 7007
          XM_005265571.1 XP_005265628.1 22 6917
          XM_005265574.3 XP_005265631.1 25 7294
          XM_006713407.1 XP_006713470.1 24 7211
          XM_006713408.1 XP_006713471.1 24 7208
          XM_006713409.1 XP_006713472.1 24 7208
          XM_006713410.1 XP_006713473.1 23 7169
          XM_006713411.1 XP_006713474.1 23 7127
          XM_006713412.1 XP_006713475.1 23 7121
          XM_006713414.3 XP_006713477.1 25 7300
          XM_011534236.1 XP_011532538.1 23 7172
          XM_011534240.2 XP_011532542.1 12 8567
          XM_011534241.2 XP_011532543.1 11 8481
          XM_017007497.1 XP_016862986.1 24 7205
          XM_017007498.1 XP_016862987.1 24 7202
          XM_017007499.1 XP_016862988.1 24 7566
          XM_017007500.1 XP_016862989.1 24 7556
          XM_017007501.1 XP_016862990.1 23 7088
          XM_017007502.1 XP_016862991.1 25 7297
          XM_017007503.1 XP_016862992.1 25 7297
          XM_017007504.1 XP_016862993.1 25 7294
          XM_017007505.1 XP_016862994.1 25 7294
          XM_017007506.1 XP_016862995.1 25 7258
          XM_017007507.1 XP_016862996.1 22 6249
          XM_017007508.1 XP_016862997.1 21 6081
          GenBank mRNA AL050184.1 - 1 241
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BQ187224.1 - 1 341 Chr. 3 - 65353863 - 65354517 [-] on Build GRCh38
          GenBankmRNA D60880.1 - 1 118 Chr. 3 - 65353944 - 65354293 [-] on Build GRCh38
          GenBankmRNA DT219440.1 - 1 597 Chr. 3 - 65354082 - 65354773 [-] on Build GRCh38
          GenBankmRNA R69913.1 - 88 Chr. - - [-] on Build
          GenBankmRNA BG426447.1 - 1 464 Chr. 3 - 65353831 - 65354640 [-] on Build GRCh38
          GenBankmRNA BF576579.1 - 323 Chr. - - [-] on Build
          GenBankmRNA CB216524.1 - 1 422 Chr. 3 - 65353964 - 65354598 [-] on Build GRCh38
          GenBankmRNA BM930454.1 - 1 51 Chr. 3 - 65353966 - 65354227 [-] on Build GRCh38

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          protein complex assembly
          cell adhesion
          cell surface receptor signaling pathway
          neuron death
          protein binding
          ATP binding
          protein C-terminus binding
          protein complex scaffold
          alpha-actinin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0