Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MAGI1 SIRNA Assays ›
Gene Symbol:
MAGI1
Gene Name:
membrane associated guanylate kinase, WW and PDZ domain containing 1
Gene Aliases:
AIP-3, AIP3, BAIAP1, BAP-1, BAP1, MAGI-1, Magi1d, TNRC19, WWP3
Chromosome Location:
Chr.3: 65353525 - 66038836 on Build GRCh38
UniGene ID:
Hs.476636
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
n288425
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
5,354.00
Your Price
Online offer:
5354.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MAGI1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
XM_005265563.1
XP_005265620.1
24
7840
XM_005265564.1
XP_005265621.1
24
7837
XM_005265565.1
XP_005265622.1
24
7807
XM_005265566.1
XP_005265623.1
24
7801
XM_005265568.1
XP_005265625.1
23
7798
XM_005265570.1
XP_005265627.1
23
7639
XM_005265571.1
XP_005265628.1
22
7549
XM_005265574.3
XP_005265631.1
25
7926
XM_006713407.1
XP_006713470.1
24
7843
XM_006713408.1
XP_006713471.1
24
7840
XM_006713409.1
XP_006713472.1
24
7840
XM_006713410.1
XP_006713473.1
23
7801
XM_006713411.1
XP_006713474.1
23
7759
XM_006713412.1
XP_006713475.1
23
7753
XM_006713414.3
XP_006713477.1
25
7932
XM_011534236.1
XP_011532538.1
23
7804
XM_011534240.2
XP_011532542.1
12
9199
XM_011534241.2
XP_011532543.1
11
9113
XM_017007497.1
XP_016862986.1
24
7837
XM_017007498.1
XP_016862987.1
24
7834
XM_017007499.1
XP_016862988.1
24
8198
XM_017007500.1
XP_016862989.1
24
8188
XM_017007501.1
XP_016862990.1
23
7720
XM_017007502.1
XP_016862991.1
25
7929
XM_017007503.1
XP_016862992.1
25
7929
XM_017007504.1
XP_016862993.1
25
7926
XM_017007505.1
XP_016862994.1
25
7926
XM_017007506.1
XP_016862995.1
25
7890
XM_017007507.1
XP_016862996.1
22
6881
XM_017007508.1
XP_016862997.1
21
6713
GenBank mRNA
AL050184.1
-
1
873
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein complex assembly
cell adhesion
cell surface receptor signaling pathway
neuron death
protein binding
ATP binding
protein C-terminus binding
protein complex scaffold
alpha-actinin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0