Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006717278.1 | XP_006717341.1 | 7 | 1136 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK021661.1 | - | 1 | 1779 | Chr. 9 - 97654435 - 97656245 [-] on Build GRCh38 |