Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIN52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011537320.2 | XP_011535622.1 | 6 | 4211 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR616131.1 | - | 1 | 872 | Chr. 14 - 74116918 - 74118235 [+] on Build GRCh38 |