Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TAF1 SIRNA Assays ›
          Gene Symbol:
          TAF1
          Gene Name:
          TATA-box binding protein associated factor 1
          Gene Aliases:
          BA2R, CCG1, CCGS, DYT3, DYT3/TAF1, KAT4, MRXS33, N-TAF1, NSCL2, OF, P250, TAF(II)250, TAF2A, TAFII-250, TAFII250, XDP
          Chromosome Location:
          Chr.X: 71366220 - 71530525 on Build GRCh38
          UniGene ID:
          Hs.158560
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n298758

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TAF1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_104387.1 40 6281
          NR_104388.1 40 6272
          NR_104389.1 39 6179
          NR_104390.1 40 6185
          NR_104391.1 41 6207
          NR_104392.1 40 6201
          NR_104393.1 41 6287
          NR_104394.1 40 6158
          NR_104395.1 40 6210
          NR_104396.1 3 1646
          GenBank mRNA AJ549245.1 - 2 1184
          AJ549246.2 - 3 1646
          AJ549247.1 - 5 971
          AJ549248.3 - 15 2106
          AJ549249.2 - 11 1643
          AJ549250.3 - 14 2004
          AJ555148.2 - 15 2010
          AJ555149.2 - 16 2032

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          general transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA-templated transcription, initiation
          transcription from RNA polymerase II promoter
          transcription initiation from RNA polymerase II promoter
          transcription elongation from RNA polymerase II promoter
          cellular response to DNA damage stimulus
          cell cycle
          viral process
          histone acetylation
          peptidyl-serine phosphorylation
          peptidyl-threonine phosphorylation
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          positive regulation of transcription from RNA polymerase II promoter
          protein autophosphorylation
          RNA polymerase II transcriptional preinitiation complex assembly
          positive regulation of transcription initiation from RNA polymerase II promoter
          regulation of signal transduction by p53 class mediator
          p53 binding
          transcription coactivator activity
          histone acetyltransferase activity
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          transcription factor binding
          TBP-class protein binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding
          lysine-acetylated histone binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-7ngt7:80/100.66.72.242:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0