Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MAPK8 SIRNA Assays ›
          Gene Symbol:
          MAPK8
          Gene Name:
          mitogen-activated protein kinase 8
          Gene Aliases:
          JNK, JNK-46, JNK1, JNK1A2, JNK21B1/2, PRKM8, SAPK1, SAPK1c
          Chromosome Location:
          Chr.10: 48306639 - 48439360 on Build GRCh38
          UniGene ID:
          Hs.138211
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n300078

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MAPK8
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001278547.1 NP_001265476.1 12 3668
          NM_001278548.1 NP_001265477.1 10 3262
          NM_001323302.1 NP_001310231.1 14 3936
          NM_001323320.1 NP_001310249.1 11 3440
          NM_001323321.1 NP_001310250.1 12 3673
          NM_001323322.1 NP_001310251.1 14 3931
          NM_001323323.1 NP_001310252.1 13 3737
          NM_001323324.1 NP_001310253.1 12 3673
          NM_001323325.1 NP_001310254.1 13 3867
          NM_001323326.1 NP_001310255.1 13 3742
          NM_001323327.1 NP_001310256.1 13 3867
          NM_001323328.1 NP_001310257.1 13 3737
          NM_001323329.1 NP_001310258.1 12 3668
          NM_001323330.1 NP_001310259.1 13 3862
          NM_001323331.1 NP_001310260.1 13 3862
          NM_139046.3 NP_620634.1 14 3936
          NM_139049.3 NP_620637.1 14 3931
          NR_136583.1 12 3669
          NR_136584.1 12 3664
          NR_136585.1 12 3669
          GenBank mRNA AK125150.1 - 1 251

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          response to stress
          JNK cascade
          JUN phosphorylation
          response to UV
          positive regulation of gene expression
          regulation of macroautophagy
          peptidyl-serine phosphorylation
          peptidyl-threonine phosphorylation
          regulation of histone deacetylation
          positive regulation of cyclase activity
          negative regulation of protein binding
          regulation of protein localization
          Fc-epsilon receptor signaling pathway
          regulation of circadian rhythm
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          rhythmic process
          regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of protein metabolic process
          cellular response to lipopolysaccharide
          cellular response to mechanical stimulus
          positive regulation of deacetylase activity
          positive regulation of protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
          protein serine/threonine kinase activity
          JUN kinase activity
          protein binding
          ATP binding
          kinase activity
          enzyme binding
          histone deacetylase regulator activity
          histone deacetylase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0