Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC15A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017018791.1 | XP_016874280.1 | 8 | 1998 | ||
XM_017018792.1 | XP_016874281.1 | 8 | 1968 | |||
XM_017018793.1 | XP_016874282.1 | 8 | 1848 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR936821.1 | - | 1 | 13 | Chr. 12 - 128793757 - 128798825 [-] on Build GRCh38 |