Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other WNT4 SIRNA Assays ›
          Gene Symbol:
          WNT4
          Gene Name:
          Wnt family member 4
          Gene Aliases:
          SERKAL, WNT-4
          Chromosome Location:
          Chr.1: 22117305 - 22143981 on Build GRCh38
          UniGene ID:
          Hs.25766
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n302020

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: WNT4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_011541597.2 XP_011539899.1 1 787
          GenBank mRNA AF086347.1 - 1 287
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA AA195302.1 - 1 286 Chr. 1 - 22141293 - 22141812 [-] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          branching involved in ureteric bud morphogenesis
          kidney development
          epithelial to mesenchymal transition
          embryonic epithelial tube formation
          liver development
          androgen biosynthetic process
          male gonad development
          female gonad development
          cellular response to starvation
          negative regulation of gene expression
          negative regulation of steroid biosynthetic process
          Wnt signaling pathway
          regulation of cell-cell adhesion
          neuron differentiation
          female sex determination
          adrenal gland development
          negative regulation of cell migration
          positive regulation of bone mineralization
          positive regulation of aldosterone biosynthetic process
          positive regulation of collagen biosynthetic process
          immature T cell proliferation in thymus
          non-canonical Wnt signaling pathway via MAPK cascade
          negative regulation of fibroblast growth factor receptor signaling pathway
          positive regulation of GTPase activity
          cell fate commitment
          negative regulation of cell differentiation
          positive regulation of osteoblast differentiation
          positive regulation of meiotic nuclear division
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          oocyte development
          smooth muscle cell differentiation
          positive regulation of stress fiber assembly
          positive regulation of focal adhesion assembly
          canonical Wnt signaling pathway
          somatotropin secreting cell differentiation
          thyroid-stimulating hormone-secreting cell differentiation
          mesenchymal to epithelial transition
          tertiary branching involved in mammary gland duct morphogenesis
          negative regulation of wound healing
          mammary gland epithelium development
          positive regulation of dermatome development
          paramesonephric duct development
          negative regulation of testicular blood vessel morphogenesis
          cellular response to transforming growth factor beta stimulus
          renal vesicle formation
          renal vesicle induction
          metanephric mesenchymal cell differentiation
          metanephric tubule formation
          metanephric nephron morphogenesis
          establishment of protein localization to plasma membrane
          negative regulation of canonical Wnt signaling pathway
          positive regulation of canonical Wnt signaling pathway
          negative regulation of male gonad development
          positive regulation of cortisol biosynthetic process
          negative regulation of testosterone biosynthetic process
          negative regulation of apoptotic signaling pathway
          transcription corepressor activity
          frizzled binding
          receptor agonist activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-qkzv8:80/100.66.72.111:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0