Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other WNT4 SIRNA Assays ›
Gene Symbol:
WNT4
Gene Name:
Wnt family member 4
Gene Aliases:
SERKAL, WNT-4
Chromosome Location:
Chr.1: 22117305 - 22143981 on Build GRCh38
UniGene ID:
Hs.25766
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
n302025
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: WNT4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
XM_011541597.2
XP_011539899.1
1
727
GenBank mRNA
AF086347.1
-
1
227
Back To Top
More Information
Panther Classification:
Molecular Function -
intercellular signal molecule
Gene Ontology Categories:
Function(s)
Process(es)
branching involved in ureteric bud morphogenesis
kidney development
epithelial to mesenchymal transition
embryonic epithelial tube formation
liver development
androgen biosynthetic process
male gonad development
female gonad development
cellular response to starvation
negative regulation of gene expression
negative regulation of steroid biosynthetic process
Wnt signaling pathway
regulation of cell-cell adhesion
neuron differentiation
female sex determination
adrenal gland development
negative regulation of cell migration
positive regulation of bone mineralization
positive regulation of aldosterone biosynthetic process
positive regulation of collagen biosynthetic process
immature T cell proliferation in thymus
non-canonical Wnt signaling pathway via MAPK cascade
negative regulation of fibroblast growth factor receptor signaling pathway
positive regulation of GTPase activity
cell fate commitment
negative regulation of cell differentiation
positive regulation of osteoblast differentiation
positive regulation of meiotic nuclear division
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
oocyte development
smooth muscle cell differentiation
positive regulation of stress fiber assembly
positive regulation of focal adhesion assembly
canonical Wnt signaling pathway
somatotropin secreting cell differentiation
thyroid-stimulating hormone-secreting cell differentiation
mesenchymal to epithelial transition
tertiary branching involved in mammary gland duct morphogenesis
negative regulation of wound healing
mammary gland epithelium development
positive regulation of dermatome development
paramesonephric duct development
negative regulation of testicular blood vessel morphogenesis
cellular response to transforming growth factor beta stimulus
renal vesicle formation
renal vesicle induction
metanephric mesenchymal cell differentiation
metanephric tubule formation
metanephric nephron morphogenesis
establishment of protein localization to plasma membrane
negative regulation of canonical Wnt signaling pathway
positive regulation of canonical Wnt signaling pathway
negative regulation of male gonad development
positive regulation of cortisol biosynthetic process
negative regulation of testosterone biosynthetic process
negative regulation of apoptotic signaling pathway
transcription corepressor activity
frizzled binding
receptor agonist activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0