Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CSF1R SIRNA Assays ›
          Gene Symbol:
          CSF1R
          Gene Name:
          colony stimulating factor 1 receptor
          Gene Aliases:
          C-FMS, CD115, CSF-1R, CSFR, FIM2, FMS, HDLS, M-CSF-R
          Chromosome Location:
          Chr.5: 150053291 - 150113372 on Build GRCh38
          UniGene ID:
          Hs.586219
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n302432

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CSF1R
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001288705.1 NP_001275634.1 1 17
          NR_109969.1 1 17
          GenBank mRNA AK304491.1 - 1 17
          DA219251.1 - 1 17
          M25786.1 - 1 112
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DC419674.1 - 1 17 Chr. 5 - 150080123 - 150086640 [-] on Build GRCh38
          GenBankmRNA AL708231.1 - 1 169 Chr. 5 - 150086394 - 150086792 [-] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein phosphorylation
          inflammatory response
          signal transduction
          transmembrane receptor protein tyrosine kinase signaling pathway
          multicellular organism development
          axon guidance
          skeletal muscle tissue development
          cell proliferation
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          regulation of cell shape
          peptidyl-tyrosine phosphorylation
          cytokine-mediated signaling pathway
          olfactory bulb development
          forebrain neuron differentiation
          hemopoiesis
          monocyte differentiation
          macrophage differentiation
          osteoclast differentiation
          positive regulation of cell migration
          ruffle organization
          cellular response to macrophage colony-stimulating factor stimulus
          macrophage colony-stimulating factor signaling pathway
          positive regulation of tyrosine phosphorylation of Stat3 protein
          negative regulation of apoptotic process
          innate immune response
          regulation of bone resorption
          cell-cell junction maintenance
          positive regulation of osteoclast differentiation
          phosphatidylinositol metabolic process
          protein autophosphorylation
          phosphatidylinositol-mediated signaling
          mammary gland duct morphogenesis
          positive regulation of protein tyrosine kinase activity
          positive regulation of ERK1 and ERK2 cascade
          cellular response to cytokine stimulus
          positive regulation of protein serine/threonine kinase activity
          positive regulation of chemokine secretion
          positive regulation of cell motility
          regulation of actin cytoskeleton reorganization
          macrophage colony-stimulating factor receptor activity
          ATP binding
          protein phosphatase binding
          cytokine binding
          protein homodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-wf5dr:80/100.66.77.5:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0