Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MZF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF147345.1 | - | 1 | 250 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA183213.1 | - | 1 | 309 | Chr. 19 - 58571623 - 58572168 [-] on Build GRCh38 |