Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAET1G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011535800.2 | XP_011534102.1 | 1 | 1763 | ||
XM_017010831.1 | XP_016866320.1 | 1 | 1763 | |||
GenBank mRNA | AK091263.1 | - | 1 | 1762 |