Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB46-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BI089231.1 | - | 3 | 377 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK000832.1 | - | 2 | 660 | Chr. 20 - 63815596 - 63817423 [+] on Build GRCh38 |