Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SMARCA5 SIRNA Assays ›
          Gene Symbol:
          SMARCA5
          Gene Name:
          SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 5
          Gene Aliases:
          ISWI, SNF2H, WCRF135, hISWI, hSNF2H
          Chromosome Location:
          Chr.4: 143513463 - 143557489 on Build GRCh38
          UniGene ID:
          Hs.558422
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n312376

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 404.00
          Your Price
          Online offer: 391.65
          391.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SMARCA5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_003601.3 NP_003592.3 24 4665
          GenBank mRNA AL157495.1 - 1 783
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DB527785.1 - 1 349 Chr. 4 - 143553882 - 143554366 [+] on Build GRCh38
          GenBankmRNA BM983819.1 - 1 254 Chr. 4 - 143553975 - 143554350 [+] on Build GRCh38
          GenBankmRNA BM473431.1 - 1 593 Chr. 4 - 143553639 - 143554364 [+] on Build GRCh38
          GenBankmRNA DB573599.1 - 1 329 Chr. 4 - 143553899 - 143554369 [+] on Build GRCh38
          GenBankmRNA BP398071.1 - 1 359 Chr. 4 - 143553876 - 143554369 [+] on Build GRCh38
          GenBankmRNA BP398546.1 - 1 358 Chr. 4 - 143553871 - 143554369 [+] on Build GRCh38
          GenBankmRNA BE540358.1 - 1 435 Chr. 4 - 143553792 - 143554372 [+] on Build GRCh38
          GenBankmRNA DB527922.1 - 1 335 Chr. 4 - 143553895 - 143554367 [+] on Build GRCh38
          GenBankmRNA DB563930.1 - 1 286 Chr. 4 - 143553944 - 143554368 [+] on Build GRCh38
          GenBankmRNA DB520295.1 - 1 313 Chr. 4 - 143553918 - 143554369 [+] on Build GRCh38
          GenBankmRNA DB554174.1 - 1 275 Chr. 4 - 143553953 - 143554369 [+] on Build GRCh38
          GenBankmRNA DB551304.1 - 1 344 Chr. 4 - 143553885 - 143554367 [+] on Build GRCh38
          GenBankmRNA BP395926.1 - 1 254 Chr. 4 - 143553976 - 143554334 [+] on Build GRCh38
          GenBankmRNA BM828760.1 - 1 457 Chr. 4 - 143553772 - 143554344 [+] on Build GRCh38
          GenBankmRNA D61483.1 - 1 261 Chr. 4 - 143553975 - 143554369 [+] on Build GRCh38
          GenBankmRNA DB528661.1 - 1 324 Chr. 4 - 143553904 - 143554369 [+] on Build GRCh38
          GenBankmRNA D81709.1 - 1 242 Chr. 4 - 143553987 - 143554342 [+] on Build GRCh38
          GenBankmRNA BM129764.1 - 1 467 Chr. 4 - 143553762 - 143554238 [+] on Build GRCh38
          GenBankmRNA AV712064.1 - 126 Chr. - - [-] on Build

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA helicase

          Gene Ontology Categories:

          Function(s) Process(es)

          chromatin silencing at rDNA
          double-strand break repair
          nucleosome assembly
          chromatin remodeling
          DNA-templated transcription, initiation
          regulation of transcription from RNA polymerase II promoter
          covalent chromatin modification
          nucleosome positioning
          CENP-A containing nucleosome assembly
          ATP-dependent chromatin remodeling
          positive regulation of gene expression, epigenetic
          positive regulation of transcription, DNA-templated
          DNA binding
          helicase activity
          protein binding
          ATP binding
          ATPase activity
          nucleosome binding
          histone binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0