Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005249807.4 | XP_005249864.1 | 7 | 5456 | ||
XM_006715752.2 | XP_006715815.1 | 8 | 5424 | |||
XM_017012414.1 | XP_016867903.1 | 7 | 5383 | |||
XM_017012415.1 | XP_016867904.1 | 8 | 5485 | |||
XM_017012417.1 | XP_016867906.1 | 8 | 3887 | |||
XM_017012418.1 | XP_016867907.1 | 7 | 3757 | |||
XM_017012421.1 | XP_016867910.1 | 8 | 5607 | |||
XM_017012422.1 | XP_016867911.1 | 8 | 5496 | |||
XM_017012424.1 | XP_016867913.1 | 7 | 5399 | |||
XM_017012430.1 | XP_016867919.1 | 8 | 3859 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK024255.1 | - | 1 | 1879 | Chr. 7 - 43628315 - 43630620 [-] on Build GRCh38 |