Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF217 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286398.2 | NP_001273327.1 | 6 | 3199 | ||
NM_152553.4 | NP_689766.1 | 9 | 2780 | |||
NR_104440.2 | 7 | 2556 | ||||
NR_136734.1 | 7 | 3320 | ||||
XM_011535494.2 | XP_011533796.1 | 7 | 3421 | |||
XM_011535496.2 | XP_011533798.1 | 6 | 3195 | |||
XM_011535500.2 | XP_011533802.1 | 7 | 2549 | |||
GenBank mRNA | AK023584.1 | - | 1 | 442 |