Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFTA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY102071.1 | - | 2, 3 | 146 | ||
BC129834.1 | - | 2, 3 | 512 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX103090.1 | - | 3, 4 | 372 | Chr. 14 - 36474749 - 36513775 [-] on Build GRCh38 |