siRNA Details
Gene Symbol: ZMAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022470.3 | NP_071915.1 | 6 | 6307 | ||
NM_152240.2 | NP_689426.1 | 7 | 6425 | |||
XM_005247706.2 | XP_005247763.1 | 6 | 6715 | |||
XM_011513073.1 | XP_011511375.1 | 7 | 6839 | |||
GenBank mRNA | BC036485.1 | - | 1 | 182 |

More Information
Panther Classification:
Molecular Function -




