Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NTRK2 SIRNA Assays ›
          Gene Symbol:
          NTRK2
          Gene Name:
          neurotrophic receptor tyrosine kinase 2
          Gene Aliases:
          GP145-TrkB, TRKB, trk-B
          Chromosome Location:
          Chr.9: 84668368 - 85027070 on Build GRCh38
          UniGene ID:
          Hs.494312
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n321595

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NTRK2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001018064.2 NP_001018074.1 17 8017
          NM_006180.4 NP_006171.2 21 8724
          XM_005252001.2 XP_005252058.1 20 8605
          XM_005252003.2 XP_005252060.1 20 8421
          XM_005252004.2 XP_005252061.1 19 8321
          XM_011518718.2 XP_011517020.1 19 8557
          XM_017014751.1 XP_016870240.1 21 9147
          XM_017014752.1 XP_016870241.1 20 8612
          XM_017014753.1 XP_016870242.1 20 9099
          XM_017014754.1 XP_016870243.1 18 8273
          GenBank mRNA BX649001.1 - 1 4181
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BM689492.1 - 1 157 Chr. 9 - 85026465 - 85026986 [+] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          vasculogenesis
          neuron migration
          positive regulation of protein phosphorylation
          inflammatory response
          neuromuscular junction development
          aging
          learning
          long-term memory
          circadian rhythm
          feeding behavior
          positive regulation of cell proliferation
          response to light stimulus
          positive regulation of gene expression
          positive regulation of neuron projection development
          response to auditory stimulus
          glutamate secretion
          positive regulation of phosphatidylinositol 3-kinase signaling
          peptidyl-tyrosine phosphorylation
          central nervous system neuron development
          cerebral cortex development
          neuron differentiation
          brain-derived neurotrophic factor receptor signaling pathway
          positive regulation of peptidyl-serine phosphorylation
          calcium-mediated signaling using intracellular calcium source
          neurotrophin signaling pathway
          mechanoreceptor differentiation
          regulation of GTPase activity
          positive regulation of MAPK cascade
          negative regulation of neuron apoptotic process
          retinal rod cell development
          protein autophosphorylation
          regulation of neurotransmitter secretion
          oligodendrocyte differentiation
          peripheral nervous system neuron development
          positive regulation of axonogenesis
          regulation of dendrite development
          regulation of protein kinase B signaling
          positive regulation of synapse assembly
          positive regulation of synaptic transmission, glutamatergic
          retina development in camera-type eye
          long-term synaptic potentiation
          cellular response to amino acid stimulus
          cellular response to tumor necrosis factor
          cellular response to brain-derived neurotrophic factor stimulus
          negative regulation of anoikis
          ATP binding
          receptor tyrosine kinase binding
          protein homodimerization activity
          neurotrophin binding
          brain-derived neurotrophic factor binding
          brain-derived neurotrophic factor-activated receptor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0