Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 企業服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NTRK2 SIRNA Assays ›
          Gene Symbol:
          NTRK2
          Gene Name:
          neurotrophic receptor tyrosine kinase 2
          Gene Aliases:
          GP145-TrkB, TRKB, trk-B
          Chromosome Location:
          Chr.9: 84668368 - 85027070 on Build GRCh38
          UniGene ID:
          Hs.494312
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n321599

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NTRK2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001018064.2 NP_001018074.1 17 7200
          NM_006180.4 NP_006171.2 21 7907
          XM_005252001.2 XP_005252058.1 20 7788
          XM_005252003.2 XP_005252060.1 20 7604
          XM_005252004.2 XP_005252061.1 19 7504
          XM_011518718.2 XP_011517020.1 19 7740
          XM_017014751.1 XP_016870240.1 21 8330
          XM_017014752.1 XP_016870241.1 20 7795
          XM_017014753.1 XP_016870242.1 20 8282
          XM_017014754.1 XP_016870243.1 18 7456
          GenBank mRNA BX649001.1 - 1 3366

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          vasculogenesis
          neuron migration
          positive regulation of protein phosphorylation
          inflammatory response
          neuromuscular junction development
          aging
          learning
          long-term memory
          circadian rhythm
          feeding behavior
          positive regulation of cell proliferation
          response to light stimulus
          positive regulation of gene expression
          positive regulation of neuron projection development
          response to auditory stimulus
          glutamate secretion
          positive regulation of phosphatidylinositol 3-kinase signaling
          peptidyl-tyrosine phosphorylation
          central nervous system neuron development
          cerebral cortex development
          neuron differentiation
          brain-derived neurotrophic factor receptor signaling pathway
          positive regulation of peptidyl-serine phosphorylation
          calcium-mediated signaling using intracellular calcium source
          neurotrophin signaling pathway
          mechanoreceptor differentiation
          regulation of GTPase activity
          positive regulation of MAPK cascade
          negative regulation of neuron apoptotic process
          retinal rod cell development
          protein autophosphorylation
          regulation of neurotransmitter secretion
          oligodendrocyte differentiation
          peripheral nervous system neuron development
          positive regulation of axonogenesis
          regulation of dendrite development
          regulation of protein kinase B signaling
          positive regulation of synapse assembly
          positive regulation of synaptic transmission, glutamatergic
          retina development in camera-type eye
          long-term synaptic potentiation
          cellular response to amino acid stimulus
          cellular response to tumor necrosis factor
          cellular response to brain-derived neurotrophic factor stimulus
          negative regulation of anoikis
          ATP binding
          receptor tyrosine kinase binding
          protein homodimerization activity
          neurotrophin binding
          brain-derived neurotrophic factor binding
          brain-derived neurotrophic factor-activated receptor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-mfr4m:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0