Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SOD2 SIRNA Assays ›
          Gene Symbol:
          SOD2
          Gene Name:
          superoxide dismutase 2, mitochondrial
          Gene Aliases:
          IPO-B, IPOB, MNSOD, MVCD6, Mn-SOD
          Chromosome Location:
          Chr.6: 159679064 - 159762529 on Build GRCh38
          UniGene ID:
          Hs.487046
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n323030

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SOD2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000636.3 NP_000627.2 5 2283
          NM_001322814.1 NP_001309743.1 4 2166
          NM_001322815.1 NP_001309744.1 4 2103
          NM_001322819.1 NP_001309748.1 5 2189
          NM_001322820.1 NP_001309749.1 5 2192
          GenBank mRNA BC041951.1 - 1 222
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BU181236.1 - 534 Chr. - - [-] on Build
          GenBankmRNA BI335208.1 - 1 396 Chr. 6 - 159680759 - 159681438 [-] on Build GRCh38
          GenBankmRNA CB994406.1 - 1 391 Chr. 6 - 159680791 - 159681412 [-] on Build GRCh38
          GenBankmRNA BG545109.1 - 1 234 Chr. 6 - 159680748 - 159681273 [-] on Build GRCh38
          GenBankmRNA BI602784.1 - 1 592 Chr. 6 - 159680989 - 159681628 [-] on Build GRCh38
          GenBankmRNA BG498902.1 - 1 292 Chr. 6 - 159680699 - 159681331 [-] on Build GRCh38
          GenBankmRNA CB998538.1 - 154 Chr. - - [-] on Build
          GenBankmRNA BQ691900.1 - 1 532 Chr. 6 - 159680770 - 159681571 [-] on Build GRCh38
          GenBankmRNA AL600953.1 - 1 553 Chr. 6 - 159681011 - 159681589 [-] on Build GRCh38
          GenBankmRNA BF247494.1 - 143 Chr. - - [-] on Build
          GenBankmRNA AA381928.1 - 1 182 Chr. 6 - 159680947 - 159681221 [-] on Build GRCh38
          GenBankmRNA CX758818.1 - 290 Chr. - - [-] on Build
          GenBankmRNA BF340777.1 - 1 501 Chr. 6 - 159680808 - 159681524 [-] on Build GRCh38
          GenBankmRNA DB265315.1 - 1 492 Chr. 6 - 159680942 - 159681531 [-] on Build GRCh38
          GenBankmRNA BI603876.1 - 1 226 Chr. 6 - 159680576 - 159681261 [-] on Build GRCh38
          GenBankmRNA HY122646.1 - 1 300 Chr. 6 - 159680856 - 159681338 [-] on Build GRCh38
          GenBankmRNA CN263464.1 - 1 430 Chr. 6 - 159680888 - 159681457 [-] on Build GRCh38
          GenBankmRNA BX506068.1 - 1 310 Chr. 6 - 159680733 - 159681349 [-] on Build GRCh38
          GenBankmRNA AF075372.1 - 1 528 Chr. 6 - 159680930 - 159681567 [-] on Build GRCh38
          GenBankmRNA DB465808.1 - 1 309 Chr. 6 - 159680922 - 159681348 [-] on Build GRCh38
          GenBankmRNA AI110873.1 - 1 528 Chr. 6 - 159680930 - 159681567 [-] on Build GRCh38
          GenBankmRNA CD640244.1 - 216 Chr. - - [-] on Build
          GenBankmRNA DB490700.1 - 1 217 Chr. 6 - 159680837 - 159681256 [-] on Build GRCh38
          GenBankmRNA HY104345.1 - 1 188 Chr. 6 - 159680785 - 159681227 [-] on Build GRCh38
          GenBankmRNA BQ881589.1 - 1 144 Chr. 6 - 159680443 - 159681183 [-] on Build GRCh38
          GenBankmRNA CD640343.1 - 1 119 Chr. 6 - 159680758 - 159681158 [-] on Build GRCh38
          GenBankmRNA BM729968.1 - 1 535 Chr. 6 - 159681004 - 159681567 [-] on Build GRCh38
          GenBankmRNA AA633017.1 - 1 157 Chr. 6 - 159680795 - 159681196 [-] on Build GRCh38
          GenBankmRNA R76245.1 - 316 Chr. - - [-] on Build
          GenBankmRNA BM459525.1 - 532 Chr. - - [-] on Build
          GenBankmRNA CD521923.1 - 1 411 Chr. 6 - 159680774 - 159681450 [-] on Build GRCh38
          GenBankmRNA BG613206.1 - 1 223 Chr. 6 - 159680755 - 159681266 [-] on Build GRCh38
          GenBankmRNA BG924293.1 - 1 296 Chr. 6 - 159679756 - 159681260 [-] on Build GRCh38
          GenBankmRNA DA814190.1 - 1 169 Chr. 6 - 159680770 - 159681208 [-] on Build GRCh38
          GenBankmRNA DB122865.1 - 1 161 Chr. 6 - 159680770 - 159681200 [-] on Build GRCh38
          GenBankmRNA BF342944.1 - 1 321 Chr. 6 - 159680583 - 159681368 [-] on Build GRCh38
          GenBankmRNA BF344034.1 - 331 Chr. - - [-] on Build
          GenBankmRNA BM470662.1 - 534 Chr. - - [-] on Build
          GenBankmRNA CB153373.1 - 1 459 Chr. 6 - 159680935 - 159681498 [-] on Build GRCh38
          GenBankmRNA CD640871.1 - 1 350 Chr. 6 - 159680773 - 159681387 [-] on Build GRCh38
          GenBankmRNA DA366639.1 - 1 354 Chr. 6 - 159680828 - 159681391 [-] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          oxidoreductase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          response to reactive oxygen species
          response to superoxide
          age-dependent response to reactive oxygen species
          release of cytochrome c from mitochondria
          vasodilation by acetylcholine involved in regulation of systemic arterial blood pressure
          regulation of transcription from RNA polymerase II promoter
          superoxide metabolic process
          regulation of blood pressure
          negative regulation of cell proliferation
          removal of superoxide radicals
          oxygen homeostasis
          negative regulation of neuron apoptotic process
          protein homotetramerization
          oxidation-reduction process
          negative regulation of oxidative stress-induced intrinsic apoptotic signaling pathway
          superoxide dismutase activity
          manganese ion binding
          identical protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0