Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195144.1 | NP_001182073.1 | 28 | 4749 | ||
XM_005246945.1 | XP_005247002.1 | 28 | 6071 | |||
XM_005246946.1 | XP_005247003.1 | 28 | 6017 | |||
XM_005246947.2 | XP_005247004.1 | 28 | 4793 | |||
XM_006712838.3 | XP_006712901.1 | 28 | 6240 | |||
XM_011512148.2 | XP_011510450.1 | 28 | 7952 | |||
XM_017005252.1 | XP_016860741.1 | 28 | 7898 | |||
GenBank mRNA | AK021632.1 | - | 1 | 587 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CB164559.1 | - | 1 | 124 | Chr. 2 - 196987569 - 196988140 [-] on Build GRCh38 |