Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STGC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY078383.1 | - | 1 | 1083 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV708531.1 | - | 204 | Chr. - - [-] on Build |