Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other YY1 SIRNA Assays ›
          Gene Symbol:
          YY1
          Gene Name:
          YY1 transcription factor
          Gene Aliases:
          DELTA, INO80S, NF-E1, UCRBP, YIN-YANG-1
          Chromosome Location:
          Chr.14: 100238765 - 100279034 on Build GRCh38
          UniGene ID:
          Hs.388927
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n337460

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: YY1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_003403.4 NP_003394.1 5 2860
          GenBank mRNA AF147416.1 - 1 358
          DA234524.1 - 1 289
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DA576611.1 - 1 265 Chr. 14 - 100278471 - 100279039 [+] on Build GRCh38
          GenBankmRNA DA904938.1 - 1 123 Chr. 14 - 100278613 - 100279168 [+] on Build GRCh38
          GenBankmRNA DA685279.1 - 1 135 Chr. 14 - 100278601 - 100279143 [+] on Build GRCh38
          GenBankmRNA BG678770.1 - 184 Chr. - - [-] on Build
          GenBankmRNA BM479654.1 - 1 161 Chr. 14 - 100278575 - 100279577 [+] on Build GRCh38
          GenBankmRNA CD244084.1 - 1 80 Chr. 14 - 100278656 - 100279483 [+] on Build GRCh38
          GenBankmRNA DA634593.1 - 1 181 Chr. 14 - 100278555 - 100279122 [+] on Build GRCh38
          GenBankmRNA DA695998.1 - 1 340 Chr. 14 - 100278396 - 100278956 [+] on Build GRCh38
          GenBankmRNA DB164168.1 - 1 205 Chr. 14 - 100278531 - 100279103 [+] on Build GRCh38
          GenBankmRNA DA691330.1 - 1 340 Chr. 14 - 100278396 - 100278962 [+] on Build GRCh38
          GenBankmRNA DA819939.1 - 1 278 Chr. 14 - 100278458 - 100279012 [+] on Build GRCh38
          GenBankmRNA DA812758.1 - 1 226 Chr. 14 - 100278510 - 100279232 [+] on Build GRCh38
          GenBankmRNA DA493496.1 - 1 340 Chr. 14 - 100278396 - 100278968 [+] on Build GRCh38
          GenBankmRNA DA507070.1 - 1 339 Chr. 14 - 100278397 - 100278962 [+] on Build GRCh38
          GenBankmRNA AW500851.1 - 1 120 Chr. 14 - 100278616 - 100279113 [+] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          double-strand break repair via homologous recombination
          transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          cellular response to DNA damage stimulus
          spermatogenesis
          response to UV-C
          negative regulation of gene expression
          cell differentiation
          cellular response to UV
          four-way junction DNA binding
          transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          enhancer sequence-specific DNA binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          transcription coactivator activity
          transcription corepressor activity
          RNA binding
          protein binding
          zinc ion binding
          transcription regulatory region DNA binding
          SMAD binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0