Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927650 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110918.1 | 3 | 269 | |||
GenBank mRNA | AL110259.1 | - | 3 | 269 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000563754.1 | - | 3 | 269 | Chr. 16 - 65141756 - 65176573 [-] on Build GRCh38 |